Learn More
Hach Company Hach Nitrate Standard, 50 mg/L NO3 (11.3 mg/L NO3-N), 500 mL
Catalog No. NC2176876 Shop All Hach Company Products
Encompass
A ready-to-use nitrate calibration standard supplying 50 mg/L NO3 (11.3 mg/L NO3-N) in a 500 mL bottle for calibration, verification, and quality control of nitrate analyzers and sensors.
  • 50 mg/L nitrate concentration (11.3 mg/L as NO3-N).
  • Ready-to-use 500 mL solution in a laboratory-grade bottle.
  • Intended for calibration and verification of nitrate sensors and analyzers.
  • Compatible with inline and benchtop nitrate measurement equipment.
  • Safety data sheet available; verify SDS before use.
This item is not returnable. View return policy
Catalog No. NC2176876 Supplier Hach Company Supplier No. LCW825
A ready-to-use nitrate calibration standard supplying 50 mg/L NO3 (11.3 mg/L NO3-N) in a 500 mL bottle for calibration, verification, and quality control of nitrate analyzers and sensors.
  • 50 mg/L nitrate concentration (11.3 mg/L as NO3-N).
  • Ready-to-use 500 mL solution in a laboratory-grade bottle.
  • Intended for calibration and verification of nitrate sensors and analyzers.
  • Compatible with inline and benchtop nitrate measurement equipment.
  • Safety data sheet available; verify SDS before use.