Learn More
AccuStandard, Inc NITRATE STD IN WATER 1000 PPM
SDP
Catalog No. NC9892057
Encompass
Anion Standard - Nitrate as Nitrogen. 100 mL. 1000 ?g/mL in 10% Hydrochloric acid.
Catalog No. NC9892057 Supplier AccuStandard, Inc Supplier No. ICNO3N10X1
Anion Standard - Nitrate as Nitrogen. 100 mL. 1000 ?g/mL in 10% Hydrochloric acid.