Learn More
CHEMetrics™ by AquaPhoenix Nitrate Test Kit - High Range Vacu-vials Instrumental Kit

Description
- K-6983 Nitrate Vacu-vials test kit is used to measure the concentration of nitrate in water with zinc reduction.
- It comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Acidifier Solution, Buffer Solution, Zinc, reaction tube with lid, syringe, 25 mL sample cup, ampoule blank, and instructions.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| No. of Tests | 30 |
| Range | 0 to 15 ppm |
| Type | Nitrate Test Kit - High Range Vacu-vials Instrumental Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.