Learn More
Hach Company Nitrate TNTplus Vial Test, HR (5-35 mg/L NO₃⁻-N), 25 Tests
Catalog No. 501998131 Shop All Hach Company Products
Encompass
Nitrate TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrate Nitrogen by the Dimethylphenol Method.USEPA approved for compliance monitoring under the Safe Drinking Water Act.
Catalog No. 50-199-8131 Supplier Hach Company Supplier No. TNT836
Nitrate TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrate Nitrogen by the Dimethylphenol Method.USEPA approved for compliance monitoring under the Safe Drinking Water Act.