Learn More
CHEMetrics™ by AquaPhoenix Nitrate Water Quality Visual Test Kit, 0 to 210 ppm as N

Description
- Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, High Range Comparator, Acidifier Solution, Buffer Solution, Zinc, reaction tube with lid, 100 uL fixed volume pipet, pipet tips, 25 mL sample cup, and instructions.
Specifications
Specifications
| Analytes | Nitrate |
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| No. of Tests | 30 |
| Range | 0 to 210 ppm |
| Type | Water Quality Visual Test Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.