Learn More
Hach Company NitraVer 5 Nitrate Reagent Accuvac Ampules, pk/25
Supplier:  Hach Company 2511025
Encompass
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach methods 8039 & 8171. Ranges: 0.1 - 10.0 or 0.3 - 30.0 mg/L NO3-N. Pack of 25 ampules.
Catalog No. 50-199-6117