Learn More
Hach Company NitraVer 5 Nitrate Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995406 Shop All Hach Company Products
Encompass
For determination of MR and HR nitrate by the NitraVer 5 cadmium reduction method. Hach methods 8039 and 8171Range: 0.1 to 10.0 or 0.3 to 30.0 mg/L as nitrate-N. Sample Size: 10 mLPack of 100 Powder Pillows
Catalog No. 50-199-5406 Supplier Hach Company Supplier No. 2106169
For determination of MR and HR nitrate by the NitraVer 5 cadmium reduction method. Hach methods 8039 and 8171Range: 0.1 to 10.0 or 0.3 to 30.0 mg/L as nitrate-N. Sample Size: 10 mLPack of 100 Powder Pillows