Learn More
Hach Company NitraVer 5 Nitrate Reagent Powder Pillows, 5 mL, pk/100
Supplier:  Hach Company 1403599
Encompass
For high range nitrate determination by the NitraVer 5 cadmium reduction method. Range: up to 50 mg/L as nitrate-N. Sample Size: 5 mL. Pack of 100 powder pillows.
Catalog No. 50-199-4896