Learn More
Hach Company NitraVer 5 Nitrate Reagent Powder Pillows, 5 mL, pk/50
Supplier:  Hach Company 1403546
Encompass
For high range nitrate determination by the NitraVer® 5 cadmium reduction method. Range: up to 50 mg/L as nitrate-N. Sample Size: 5 mL. Pack of 50 powder pillows. For use with the Nitrate Color Cube Test Kit.
Catalog No. 50-199-4895