Learn More
Hach Company NitraVer 6 Nitrate Reagent Powder Pillows, 5 mL, pk/100
Supplier:  Hach Company 1412099
Encompass
For determination of low range nitrate by the cadmium reduction method. For use with Hach visual test kits. Use with NitriVer 3. Sample Size: 5 mL.
Catalog No. 50-199-4933