Learn More
Hach Company NitraVer 6 Nitrate Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995416 Shop All Hach Company Products
Encompass
For determination of low range nitrate by the NitraVer 6 cadmium reduction method. Range: 0.01 to 0.50 mg/L as nitrate-N. Use with NitriVer 3. Sample Size: 10 mL. Pack of 100 Powder Pillows
Catalog No. 50-199-5416 Supplier Hach Company Supplier No. 2107249
For determination of low range nitrate by the NitraVer 6 cadmium reduction method. Range: 0.01 to 0.50 mg/L as nitrate-N. Use with NitriVer 3. Sample Size: 10 mL. Pack of 100 Powder Pillows