Learn More
Hach Company NitraVer 6 Nitrate Reagent Powder Pillows, 30 mL, pk/100
Catalog No. 501994932 Shop All Hach Company Products
Encompass
For determination of low range nitrate by the cadmium reduction method. Range: up to 0.50 mg/L as nitrate-N. Use with NitriVer 3. Sample Size: 30 mL.
Catalog No. 50-199-4932 Supplier Hach Company Supplier No. 1411999
For determination of low range nitrate by the cadmium reduction method. Range: up to 0.50 mg/L as nitrate-N. Use with NitriVer 3. Sample Size: 30 mL.