Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrazine Yellow

Catalog No. aaj6028114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6028114 25 g
AAJ6028103 1 g
AAJ6028106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6028114 Supplier Thermo Scientific Chemicals Supplier No. J6028114
Only null left
Add to Cart

pH indicator | CAS: 5423-07-4 | C16H8N4Na2O11S2 | 542.36 g/mol

Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar's passivation film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar′s passivation film.

Solubility
Dissolve in water at 1mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5423-07-4
MDL Number MFCD00003938
Synonym C.I. 14890; Delta Yellow

Specifications

Molecular Formula C5H5N3O2
Solubility Information Dissolve in water at 1mg/ml
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
SMILES C1=CN=C(C(=N1)C(=O)O)N
IUPAC Name 3-aminopyrazine-2-carboxylic acid
Molecular Weight (g/mol) 139.11
PubChem CID 72656
Formula Weight 542.36
Color Brown to Orange
Quantity 25 g
Physical Form Powder
Chemical Name or Material Nitrazine Yellow
Show More Show Less
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.