Learn More
Cayman Chemical Nitrendipine, Purity: ≥95%, CAS: 39562-70-4, 10 mg
SDP
Catalog No. 502966601
Encompass_Preferred
A dihydropyridine used as an antihypertensive agent due to its ability to block L-type and T-type calcium channels; also binds to adenosine A1, A2A, and A3 receptors with Ki values of 8.96, 23.0, and 8.3 µM, respectively
Catalog No. 50-296-6601 Supplier Cayman Chemical Supplier No. 1754910 MG
A dihydropyridine used as an antihypertensive agent due to its ability to block L-type and T-type calcium channels; also binds to adenosine A1, A2A, and A3 receptors with Ki values of 8.96, 23.0, and 8.3 µM, respectively