Learn More
Medchemexpress LLC HY-B0424 100mg Medchemexpress, Nitrendipine CAS:39562-70-4 Purity:>98%
SDP
Catalog No. 502082529
Encompass_Preferred
Medchemexpress, HY-B0424 100mg Nitrendipine CAS:39562-70-4 Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2529 Supplier Medchemexpress LLC Supplier No. HYB0424100MG
Medchemexpress, HY-B0424 100mg Nitrendipine CAS:39562-70-4 Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.