Learn More
Apexbio Technology LLC Nitrendipine 39562-70-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061512
Encompass_Preferred
Nitrendipine (CAS number 39562-70-4) is a dihydropyridine derivative acting as a calcium channel antagonist that selectively inhibits L-type calcium channels By blocking calcium ion influx into vascular smooth muscle cells nitrendipine induces vasodilation and reduces peripheral vascular resistance thereby lowering arterial blood pressure Due to its vascular-selective calcium antagonism it serves as a pharmacological tool in cardiovascular research particularly for studying mechanisms of hypertension and calcium-dependent signaling pathways
Catalog No. 50-000-61512 Supplier Apexbio Technology LLC Supplier No. B13935.1
Nitrendipine (CAS number 39562-70-4) is a dihydropyridine derivative acting as a calcium channel antagonist that selectively inhibits L-type calcium channels By blocking calcium ion influx into vascular smooth muscle cells nitrendipine induces vasodilation and reduces peripheral vascular resistance thereby lowering arterial blood pressure Due to its vascular-selective calcium antagonism it serves as a pharmacological tool in cardiovascular research particularly for studying mechanisms of hypertension and calcium-dependent signaling pathways