Learn More
American Radiolabeled Chemicals Inc NITRENDIPINE 3H 25UC
SDP
Catalog No. 50353031
Encompass
Nitrendipine-5-methyl-3H 25uCi in ethanol, 1 mCi/ml, 70-87 Ci/mmol, M.W. 360.3, Shipped in Dry Ice
Catalog No. 50-353-031 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 195025UC
Nitrendipine-5-methyl-3H 25uCi in ethanol, 1 mCi/ml, 70-87 Ci/mmol, M.W. 360.3, Shipped in Dry Ice