Learn More
Medchemexpress LLC Nitrendipine-d5 | 2469554-26-3 | 99.77% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0424S5MG
Encompass_Preferred
Nitrendipine-d5 is a deuterium-labeled version of Nitrendipine, an orally active analog of Nifedipine. It acts as a dihydropyridine calcium channel blocker, shown to induce apoptosis and exhibit antihypertensive effects. Nitrendipine can also block alcohol and morphine withdrawal symptoms, reduce right ventricular hypertrophy and pulmonary vascular changes caused by intermittent hypoxia, and possess anticancer effects on neuroblastoma.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08909