Learn More
Medchemexpress LLC Nitrendipine-d5 | 2469554-26-3 | 99.8% | 1 MG
SDP
Catalog No. 5000408907
Encompass_Preferred
Nitrendipine-d5 is a deuterium-labeled version of Nitrendipine, an orally active analog of Nifedipine that functions as a dihydropyridine calcium channel blocker. It has been shown to induce apoptosis and possess antihypertensive effects. Additionally, it can block alcohol and morphine withdrawal symptoms, reduce right ventricular hypertrophy, and alleviate pulmonary vascular changes caused by intermittent hypoxia. It also exhibits anticancer effects on neuroblastoma.
  • Application as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08907 Supplier Medchemexpress LLC Supplier No. HYB0424S1MG
Nitrendipine-d5 is a deuterium-labeled version of Nitrendipine, an orally active analog of Nifedipine that functions as a dihydropyridine calcium channel blocker. It has been shown to induce apoptosis and possess antihypertensive effects. Additionally, it can block alcohol and morphine withdrawal symptoms, reduce right ventricular hypertrophy, and alleviate pulmonary vascular changes caused by intermittent hypoxia. It also exhibits anticancer effects on neuroblastoma.
  • Application as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs