Learn More
Medchemexpress LLC Nitrendipine-d5 | 2469554-26-3 | 99.8% | 100 UG
SDP
Catalog No. 5000405278
Encompass_Preferred
Nitrendipine-d5 is a deuterium-labeled analog of Nifedipine and a dihydropyridine calcium channel blocker. It has been studied for its ability to induce apoptosis, demonstrate antihypertensive and anticancer effects, and block withdrawal symptoms.
  • Functions as a dihydropyridine calcium channel blocker
  • Induces apoptosis
  • Exhibits antihypertensive effects
  • Blocks alcohol and morphine withdrawal symptoms
  • Reduces right ventricular hypertrophy and pulmonary vascular changes
  • Demonstrates anticancer effects on neuroblastoma
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05278 Supplier Medchemexpress LLC Supplier No. HYB0424S100UG
Nitrendipine-d5 is a deuterium-labeled analog of Nifedipine and a dihydropyridine calcium channel blocker. It has been studied for its ability to induce apoptosis, demonstrate antihypertensive and anticancer effects, and block withdrawal symptoms.
  • Functions as a dihydropyridine calcium channel blocker
  • Induces apoptosis
  • Exhibits antihypertensive effects
  • Blocks alcohol and morphine withdrawal symptoms
  • Reduces right ventricular hypertrophy and pulmonary vascular changes
  • Demonstrates anticancer effects on neuroblastoma
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS