Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitrendipine-d5 | 2469554-26-3 | 99.8% | 365.39 | C18H15D5N2O6 | 10 MG
SDP

Catalog No. 5000298912
Encompass_Preferred

Nitrendipine-d5 is the deuterium-labeled analog of nitrendipine used as a stable-isotope internal standard and research reagent for analytical and pharmacological studies. It functions as a dihydropyridine calcium channel blocker and is supplied as a 10 mg solid for laboratory use.

  • Deuterium-labeled internal standard for analytical assays.
  • Dihydropyridine calcium channel blocker used in pharmacology studies.
  • High isotopic and chemical purity (99.8%).
  • Molecular weight 365.39; formula C18H15D5N2O6.
  • Off-white to yellow solid appearance.
  • Supplied as a 10 mg quantity; store protected from light at 4°C.
  • Shelf life in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-98912 Supplier Medchemexpress LLC Supplier No. HYB0424S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrendipine-d5 is the deuterium-labeled analog of nitrendipine used as a stable-isotope internal standard and research reagent for analytical and pharmacological studies. It functions as a dihydropyridine calcium channel blocker and is supplied as a 10 mg solid for laboratory use.

  • Deuterium-labeled internal standard for analytical assays.
  • Dihydropyridine calcium channel blocker used in pharmacology studies.
  • High isotopic and chemical purity (99.8%).
  • Molecular weight 365.39; formula C18H15D5N2O6.
  • Off-white to yellow solid appearance.
  • Supplied as a 10 mg quantity; store protected from light at 4°C.
  • Shelf life in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.