Learn More
Selleck Chemical LLC Nitrendipine 50mg 39562-70-4 Bayotensin
SDP
Supplier:  Selleck Chemical LLC S249150MG
Encompass_Preferred
Nitrendipine (Bayotensin) is a dihydropyridine calcium channel blocker with an IC50 of 95 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-740-7