Learn More
Petroleum Analyzer Corporation Sample box Nitrogen - Sulfur Calibration Standard 25 mg/L in i-Octane (5x1.5 ml)
SDP
Catalog No. NC2152019
Encompass
Sample box Nitrogen - Sulfur Calibration Standard 25 mg/L in i-Octane (5x1.5 ml)
This item is not returnable. View return policy
Catalog No. NC2152019 Supplier Petroleum Analyzer Corporation Supplier No. 20001.711
Sample box Nitrogen - Sulfur Calibration Standard 25 mg/L in i-Octane (5x1.5 ml)