Learn More
Hach Company Nitrogen (Total) TNTplus Vial Test, UHR (20-100 mg/L N), 25 Tests
Catalog No. 501998124 Shop All Hach Company Products
Encompass
Nitrogen TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Total Nitrogen by the Persulfate Digestion method.
Catalog No. 50-199-8124 Supplier Hach Company Supplier No. TNT828
Nitrogen TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Total Nitrogen by the Persulfate Digestion method.