Learn More
MSC North Chemical-Resistant Gloves, Nitrile, Supported, Size 8, 12 in., 6/Pk.
Catalog No. 5000508395
Encompass
These supported nitrile gloves feature a durable nitrile coating over a seamless polyester and cotton interlock liner, providing robust protection against cuts, snags, abrasions, and punctures. Designed for demanding tasks including chemical handling, automotive assembly, and food processing, these gloves are treated with anti-bacterial and anti-mildew agents to maintain hygiene. The roughed finish on the palm and fingers ensures a secure grip in dry, wet, or oily conditions.
  • Roughed nitrile coating on palm and fingers for enhanced grip
  • Thermal protection ranging from -4°F to 392°F
  • Compliant with USDA and FDA 21 CFR regulations for food processing
  • Size 8 medium fit with 12 in. overall length
  • Slip-on design with a pinked cuff
  • Resistant to grease, caustic substances, solvents, and tears
Catalog No. 50-005-08395 Supplier MSC Supplier No. 51376895
These supported nitrile gloves feature a durable nitrile coating over a seamless polyester and cotton interlock liner, providing robust protection against cuts, snags, abrasions, and punctures. Designed for demanding tasks including chemical handling, automotive assembly, and food processing, these gloves are treated with anti-bacterial and anti-mildew agents to maintain hygiene. The roughed finish on the palm and fingers ensures a secure grip in dry, wet, or oily conditions.
  • Roughed nitrile coating on palm and fingers for enhanced grip
  • Thermal protection ranging from -4°F to 392°F
  • Compliant with USDA and FDA 21 CFR regulations for food processing
  • Size 8 medium fit with 12 in. overall length
  • Slip-on design with a pinked cuff
  • Resistant to grease, caustic substances, solvents, and tears