Learn More
Honeywell™ Nitri-Knit™ Supported Nitrile Gloves With Insulated Liner
Description
- High nitrile content offers resistance to cuts, snags, abrasions and punctures
- Ideal for petrochemical refining, degreasing operations, automotive assembly and chemical handling
- Free of latex proteins which can cause allergic reactions
- Will not swell, sag or weaken from fat absorption
- True dip coated glove has a permanent bond of nitrile to inner-liner
Specifications
Specifications
| Certifications/Compliance | 21 CFR 170-199, ASTM D3389-05, 29 CFR 1915.157 |
| Certifications/Compliance (Europe) | EN 420, EN 374 |
| Chlorinated | No |
| Color | Blue |
| Latex Free | Yes |
| Material | Nitrile |
| Size | 8 |
| EN388 Standard | EN 388 |
| EN 421 | No |
| For Use With (Application) | Automotive assembly, painting, chemical handling, processing, degreasing, dip tank cleanup, fishing industry, food processing, general maintenance work, printing cleanup, refinery (general), Solvents (general) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.