Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 1% (v/v), Ricca Chemical
SDP

Catalog No. R53130001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R53130001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R53130001A Supplier Ricca Chemical Company Supplier No. R53130001A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
CAS Min % 97.0, 1.45
CAS Max % 100.0, 1.51
Concentration 1% (v/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Percent Purity 1% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.