Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 10% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18611990 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-990 1 L Amber Glass Bottle
18-611-992 2.5 L Poly-Coated Glass Bottle
18-611-991 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-990 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1651LT

Due to product restrictions, please to purchase or view availability for this product.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity 10% ±0.5%
Grade Ungraded
Chemical Name or Material Nitric Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.