Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 10% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18611991 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-991 55 gal. Drum
18-611-990 1 L Amber Glass Bottle
18-611-992 2.5 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-991 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N16555GLBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5

Specifications

Packaging Drum
Quantity 55 gal.
Percent Purity 10% ±0.5%
Grade Ungraded
Chemical Name or Material Nitric Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.