Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 10% w/v, Reagents

Catalog No. 01334645
Change view
Click to view available options
Quantity: 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-645 Quantity: 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-645 Supplier Reagents Holdings Llc Supplier No. CN1043002.5D1
 Restricted Item
Add to Cart
Add to Cart

Nitric Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.0533
Quantity 2.5 L
Physical Form Liquid
Percent Purity 10% w/v
Grade Reagent
Chemical Name or Material Nitric Acid, 10% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.