Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 20% (v/v), Ricca Chemical
SDP

Catalog No. R532600025D Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R532600025D 2.5 L Safety-coated Glass Bottle
5326-16 500 mL Amber Glass Bottle
5326-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R532600025D Supplier Ricca Chemical Company Supplier No. R53260002.5D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance of Solution Colorless to Light Yellow Liquid
Color Colorless to Yellow
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Packaging Safety-coated Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 2.5 L
Synonym hydrogen nitrate
Percent Purity 20% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid, 20% (v/v)
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.