Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions NITRIC ACID 30%, V/V 500ML
SDP

Catalog No. NC0881611
Encompass_Preferred

Nitric Acid 30% v/v Solution (500 ml)

Catalog No. NC0881611 Supplier Aqua Solutions Supplier No. 6476500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitric Acid 30% v/v Solution (500 ml)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.