Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 45% (v/v), Ricca Chemical
SDP

Catalog No. R534500025D Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R534500025D 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R534500025D Supplier Ricca Chemical Company Supplier No. R53450002.5D
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 63.01 g/mol
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Appearance of Solution Colorless to Light Yellow Liquid
Concentration 44.7 to 45.3 N
CAS Max % 63.62, 36.38
Packaging Safety-coated Glass Bottle
Quantity 2.5 L
Percent Purity 45% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.