Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Nitric Acid 5N 500Ml P

Catalog No. 504400251
Encompass_Preferred

Nitric Acid 5.0N-Bottle 16Oz Plast

Catalog No. 50-440-0251 Supplier LaMarKa Chemical Company Supplier No. 0000106916748337
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitric Acid 5.0N-Bottle 16Oz Plast

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.