Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 60.0%, Ultrapur, MilliporeSigma™

Catalog No. M1015180250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Fiber Carton
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1015180250 250 mL Plastic Bottle
M1015180500 500 mL Fiber Carton
M1015181000 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1015180250 Supplier MilliporeSigma Supplier No. 1015180250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH < 1 (H2O, 20°C)
Melting Point −22°C
Boiling Point 120°C (1013 hPa)
Density 1.37 g/cm3 (at 20°C)
Packaging Plastic Bottle
Assay Percent Range ≥ 60.0%
Quantity 250 mL
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.