Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 65%, Suprapur™, MilliporeSigma™

Catalog No. M1004411000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1004411000 1 L Glass Bottle
M1004410250 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1004411000 Supplier MilliporeSigma Supplier No. 1004411000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Density 1.39g/cm3 (20°C)
Packaging Glass Bottle
Vapor Pressure 9.4 hPa (20°C)
Assay Percent Range 65%
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble (20°C)
Grade Suprapur
Chemical Name or Material Nitric Acid, Suprapur™
Hazard Category SignalWord: Danger
Recommended Storage Store below +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.