Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid 70%, Trace Metal Also meets ACS specifications, Reagents

Catalog No. 01336827
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-827 2.5 L Glass Bottle
01-336-829 2.5 L Poly Bottle
01-336-828 2.5 L Safety Coated Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-827 Supplier Reagents Holdings Llc Supplier No. C51016502.5D1
 Restricted Item
Add to Cart
Add to Cart

Nitric Acid 70%, Trace Metal Also meets ACS specifications, Trace Metal, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate, Aqua fortis
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH <1
Color Colorless to Slight Yellow
Boiling Point 120.5°C
Packaging Glass Bottle
Specific Gravity 1.5027
Quantity 2.5 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Nitric Acid 70%, Trace Metal Also meets ACS specifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.