Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 70% (w/w), Trace Metals Grade, Ricca Chemical
SDP

Catalog No. RABN0020 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RABN0020 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RABN0020 Supplier Ricca Chemical Company Supplier No. RABN00202.5D1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitric Acid, 70% (w/w), Trace Metals Grade, Ricca Chemical

  • Nitric Acid, 70% (w/w), Trace Metals Grade

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 63.01 g/mol
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Appearance Clear, Colorless to Light Yellow
Melting Point -41.6°C
Boiling Point 82.6°C
Packaging Glass
CAS Max % 70%, 30%
CAS Min % 70%, 30%
Assay Percent Range 68 to 70% (w/w)
Quantity 2.5 L
Percent Purity 68 to 70% W/W
Grade ICP-OES
Chemical Name or Material Nitric Acid
Show More Show Less
ShelfLife 24 Months
Recommended Storage Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.