Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, 90+%, ACS reagent, fuming, Thermo Scientific Chemicals

Catalog No. AC270620010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270620010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC270620010 Supplier Thermo Scientific Chemicals Supplier No. 270620010
  • $449.00
    $402.65 / Each of 1

    Save $46.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Blue to Blue-Green
Melting Point -40.0°C
Boiling Point 84.0°C
Density 1.5110g/mL
Heavy Metals (as Pb) 5ppm max.
Packaging Glass bottle
Residue after Ignition 0.002 % max.
Specific Gravity 1.511
Assay Percent Range 90+%
Quantity 1 L
Viscosity 0.746 mPa.s (25°C)
Physical Form Crystals or Crystalline Powder
Linear Formula HNO3
Merck Index 15, 6663
Solubility Information Solubility in water: miscible.
Formula Weight 63.01
Percent Purity ≥90%
Grade ACS Reagent
Chemical Name or Material Nitric acid, For analysis ACS, Fuming
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer. May be corrosive to metals. Causes severe skin burns and eye damage. Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Take any precaution to avoid mixing with combustibles.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.