Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid ACS AR, Macron Fine Chemicals™

Catalog No. MK2704500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass
SAFEMOR
Grade:
ACS
AR Select ACS
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Grade
MK-2704-500 500 mL Glass ACS
MK-1409-500 500 mL SAFEMOR ACS
MK-1409-212 2.5 L SAFEMOR ACS
MK-6623-212 2.5 L SAFEMOR AR Select ACS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MK-2704-500 Supplier Avantor Supplier No. 270414
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Glass
Quantity 500 mL
Formula Weight 63.01
Grade ACS
Chemical Name or Material Nitric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.