Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nitric acid, ACS, fuming, 90%

Catalog No. AA33261K4
Change view
Click to view available options
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA33261K4 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33261K4 Supplier Thermo Scientific™ Supplier No. 33261K4

missing translation for 'validMainframeMsg'

Specifications

Melting Point ca -50°
CAS 7697-37-2
Boiling Point 85°
Density 1.5
Quantity 2.5 L
Molecular Formula HNO{3}
Physical Form Liquid
MDL Number MFCD00011349
UN Number UN2032
Merck Index 14,6577
Formula Weight 63.01
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.