Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals

Catalog No. AC424000250
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Plastic bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424000250 25 mL Glass Bottle
AC424005000 500 mL Glass Bottle
AC424005001 500 mL Plastic bottle
AC424000025 2.5 L Glass Bottle
AC424000026 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC424000250 Supplier Thermo Scientific Chemicals Supplier No. 424000250
 Restricted Item
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Melting Point -42.0°C
Boiling Point 122.0°C
CAS Min % 68.0, 30.0
CAS Max % 70.0, 32.0
Density 1.4000g/mL
Heavy Metals (as Pb) 0.2ppm max.
Packaging Glass Bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.4
Quantity 25 mL
Physical Form Liquid Free From Suspended Matter or Sediment
Linear Formula HNO3
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Solubility Information Solubility in water: >100 g/100mL (20°C).
Formula Weight 63.01
Percent Purity 68.0 to 70.0%
Grade ACS Reagent
Chemical Name or Material Nitric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep/Store away from clothing/ combustible materials.
Recommended Storage this material may darken during storage due to

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.