Learn More
Sigma Aldrich Fine Chemicals Biosciences Nitric acid ACS reagent 70
Catalog No. 502902883
Encompass
Nitric acid (HNO3) is a highly corrosive mineral acid. It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them. The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight Raman spectrum solubility morphology and alignment examinations.
Catalog No. 50-290-2883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4380734X2.5L
 Restricted Item
Add to Cart
Add to Cart
Nitric acid (HNO3) is a highly corrosive mineral acid. It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them. The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight Raman spectrum solubility morphology and alignment examinations.