Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, ACS Reagent, 70%, Solstice

Catalog No. 60046879 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.2 L
2.5 L
Packaging:
Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-879 2.2 L Poly Bottle
60-046-878 100 mL Glass Bottle
60-010-90 500 mL Glass Bottle
60-010-88 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-879 Supplier Solstice Supplier No. 4380732.2LP

missing translation for 'validMainframeMsg'

ACS Reagent, 70%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH Acidic
Melting Point -38°C
Boiling Point 122°C (1013hPa)
Density 1.413g/mL at 20°C
Odor Stinging
Packaging Poly Bottle
Vapor Pressure 8mmHg (20°C)
Quantity 2.2 L
Physical Form Liquid
UN Number UN2031
Formula Weight 63.01g/mol
Percent Purity 68.0 to 70.0%
Chemical Name or Material Nitric acid
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
DOTInformation class 8 (5.1) - PG 2 - IATA PAX forbidden - Nitric acid ≥65% - ≤70%
RTECSNumber 231-714-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.