Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, ACS Reagent Grade, 70%, Ricca Chemical
SDP

Catalog No. RABN001025D Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABN001025D 2.5 L Glass Bottle
RABN0010500 500 mL Glass Bottle
RABN00101C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. RABN001025D Supplier Ricca Chemical Company Supplier No. RABN00102.5D1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass Bottle
Quantity 2.5 L
Physical Form Liquid
Percent Purity 68 to 70%
Grade ACS
Chemical Name or Material Nitric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.