Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 62% (w/w), Ricca Chemical
SDP

Catalog No. R5352620500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5352620500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5352620500 Supplier Ricca Chemical Company Supplier No. R5352620500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Weight (g/mol) 63.01 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Appearance Clear colorless to pale yellow liquid
Odor Slightly pungent to pungent
Packaging Amber Glass Bottle
Assay Percent Range 62% (w/w)
Quantity 500 mL
Physical Form Liquid
Solubility Information Solubility in Water: Infinite
Percent Purity 62% W/W
Grade Laboratory
Chemical Name or Material Nitric Acid
Recommended Storage Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.