Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, Concentrate, 0.1 M HNO3 in Water (0.1 N), Eluent, Concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111020293 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0293 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0293 Supplier MilliporeSigma Supelco Supplier No. 163551L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011349
Synonym Nitric acid solution

Specifications

CAS 7697-37-2
Linear Formula HNO3
Quantity 1 L
Formula Weight 63.01
Concentration or Composition (by Analyte or Components) 0.1 M HNO 3 aq. soln. (0.1 N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.