Learn More
GFS Chemicals Inc NITRIC ACID, VERITAS DOUBLE DISTILLED | 500 ML | GFS CHEMICALS, INC. Item #: 53202 | Bulk #: 621
SDP
Catalog No. NC9735314
Encompass_Preferred
NITRIC ACID, VERITAS DOUBLE DISTILLED | Chemical Formula : HNO!3 | CAS # : 7697-37-2 | Specifications : Ag (ppt), Al (ppt), Arsenic 0.0000001 % , As (ppt), Assay 68.0 % - Min, Au (ppt), B (ppt), Ba (ppt), Be (ppt), Bi (ppt), Ca (ppt), Cd (ppt), Ce (ppt), Chloride 0.00001 % , Co (ppt), Cr (ppt), Cs (ppt), Cu (ppt), Dy (ppt), Er (ppt), Eu (ppt), Fe (ppt), Ga (ppt), Gd (ppt), Ge (ppt), Heavy Metals 0.000001 % , Hf (ppt), Hg (ppt), Ho (ppt), In (ppt), Ir (ppt), Iron 0.000001 % , K (ppt), La (ppt), Li (ppt), Lu (ppt), Mg (ppt), Mn (ppt), Mo (ppt), Na (ppt), Nb (ppt), Nd (ppt), Ni (ppt), Os (ppt), P (ppt), Pb (ppt), Pd (ppt), Pr (ppt), Pt (ppt), Rb (ppt), Re (ppt), Residue after ignition 0.00001 % , Rh (ppt), Ru (ppt), Sb (ppt), Sc (ppt), Se (ppt), Si (ppt), Sm (ppt), Sn (ppt), Sr (ppt), Sulfate 0.00001 % , Ta (ppt), Tb (ppt), Te (ppt), Th (ppt), Ti (ppt), Tl (ppt), Tm (ppt), U (ppt), V (ppt), W (ppt), Y (ppt), Yb (ppt), Zn (ppt), Zr (ppt)
Catalog No. NC9735314 Supplier GFS Chemicals Inc Supplier No. 53202
NITRIC ACID, VERITAS DOUBLE DISTILLED | Chemical Formula : HNO!3 | CAS # : 7697-37-2 | Specifications : Ag (ppt), Al (ppt), Arsenic 0.0000001 % , As (ppt), Assay 68.0 % - Min, Au (ppt), B (ppt), Ba (ppt), Be (ppt), Bi (ppt), Ca (ppt), Cd (ppt), Ce (ppt), Chloride 0.00001 % , Co (ppt), Cr (ppt), Cs (ppt), Cu (ppt), Dy (ppt), Er (ppt), Eu (ppt), Fe (ppt), Ga (ppt), Gd (ppt), Ge (ppt), Heavy Metals 0.000001 % , Hf (ppt), Hg (ppt), Ho (ppt), In (ppt), Ir (ppt), Iron 0.000001 % , K (ppt), La (ppt), Li (ppt), Lu (ppt), Mg (ppt), Mn (ppt), Mo (ppt), Na (ppt), Nb (ppt), Nd (ppt), Ni (ppt), Os (ppt), P (ppt), Pb (ppt), Pd (ppt), Pr (ppt), Pt (ppt), Rb (ppt), Re (ppt), Residue after ignition 0.00001 % , Rh (ppt), Ru (ppt), Sb (ppt), Sc (ppt), Se (ppt), Si (ppt), Sm (ppt), Sn (ppt), Sr (ppt), Sulfate 0.00001 % , Ta (ppt), Tb (ppt), Te (ppt), Th (ppt), Ti (ppt), Tl (ppt), Tm (ppt), U (ppt), V (ppt), W (ppt), Y (ppt), Yb (ppt), Zn (ppt), Zr (ppt)