Learn More
IMEB, Inc Tek-Select Nitric Acid Decalcifier, 32 oz Bottle, AFIP Formulation That Blends Electronic Grade Nitric Acid with Reagent Grade Distilled Water, Widely Used Method of Rapid Decalcification Of Bone
SDP
Catalog No. 501689480
Encompass_Preferred
Tek-Select Nitric Acid Decalcifier, 32 oz Bottle. Nitric Acid Decalcifier is an AFIP formulation that blends electronic grade Nitric Acid with reagent grade distilled water. The most widely used method of rapid decalcification of bone in pathology.
Catalog No. 50-168-9480 Supplier IMEB, Inc Supplier No. NA32
Tek-Select Nitric Acid Decalcifier, 32 oz Bottle. Nitric Acid Decalcifier is an AFIP formulation that blends electronic grade Nitric Acid with reagent grade distilled water. The most widely used method of rapid decalcification of bone in pathology.