Learn More
MSC NITRIC ACID DETECTOR
Catalog No. 5000803551
Encompass
1-20PPM SNGL TB 10PK NITRIC ACID DETECTOR
Catalog No. 50-008-03551 Supplier MSC Supplier No. 47551916
1-20PPM SNGL TB 10PK NITRIC ACID DETECTOR